Product Details

SNP ID
rs138739351
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:50319033 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCCCCAAGCACATTCCTTCCTCA[C/T]GAGTCCCAGGTCCAGCTTTATTACC
Phenotype
MIM: 600385
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ADCY7 PubMed Links

Gene Details

Gene
ADCY7
Gene Name
adenylate cyclase 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001114.4 2380 Intron NP_001105.1
NM_001286057.1 2380 Intron NP_001272986.1
XM_011522835.2 2380 Intron XP_011521137.1
XM_011522836.2 2380 Intron XP_011521138.1
XM_011522837.2 2380 Intron XP_011521139.1
XM_011522838.2 2380 Intron XP_011521140.1
XM_011522839.2 2380 Intron XP_011521141.1
XM_011522840.2 2380 Intron XP_011521142.1
XM_011522842.2 2380 Intron XP_011521144.1
XM_017022895.1 2380 Intron XP_016878384.1
XM_017022896.1 2380 Intron XP_016878385.1
XM_017022897.1 2380 Intron XP_016878386.1
XM_017022898.1 2380 Intron XP_016878387.1
XM_017022899.1 2380 Intron XP_016878388.1
Gene
BRD7
Gene Name
bromodomain containing 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001173984.2 2380 Intron NP_001167455.1
NM_013263.4 2380 Intron NP_037395.2
XM_011523046.2 2380 Intron XP_011521348.1
XM_011523047.2 2380 UTR 3 XP_011521349.1
XM_011523048.2 2380 Intron XP_011521350.1
XM_011523049.2 2380 Intron XP_011521351.1
XM_011523050.2 2380 UTR 3 XP_011521352.1
XM_017023179.1 2380 Intron XP_016878668.1
XM_017023180.1 2380 Intron XP_016878669.1
XM_017023181.1 2380 UTR 3 XP_016878670.1

View Full Product Details