Product Details
- SNP ID
-
rs139282045
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:1706348 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CGGCGGCGGTGGCCGCGATGATGGA[A/G]ATCCAGATGGACGAGGGCGGCGGCG
- Phenotype
-
MIM: 605431
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
HN1L
PubMed Links
Gene Details
- Gene
- HN1L
- Gene Name
- hematological and neurological expressed 1-like
There are no transcripts associated with this gene.
- Gene
- MAPK8IP3
- Gene Name
- mitogen-activated protein kinase 8 interacting protein 3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001040439.1 |
166 |
Silent Mutation |
GAA,GAG |
E3E |
NP_001035529.1 |
NM_001318852.1 |
166 |
Silent Mutation |
GAA,GAG |
E3E |
NP_001305781.1 |
NM_015133.4 |
166 |
Silent Mutation |
GAA,GAG |
E3E |
NP_055948.2 |
XM_005255190.2 |
166 |
Silent Mutation |
GAA,GAG |
E3E |
XP_005255247.1 |
XM_006720869.2 |
166 |
Intron |
|
|
XP_006720932.1 |
XM_011522429.2 |
166 |
Silent Mutation |
GAA,GAG |
E3E |
XP_011520731.1 |
XM_011522430.2 |
166 |
Silent Mutation |
GAA,GAG |
E3E |
XP_011520732.1 |
XM_011522431.2 |
166 |
Silent Mutation |
GAA,GAG |
E3E |
XP_011520733.1 |
XM_011522432.2 |
166 |
Silent Mutation |
GAA,GAG |
E3E |
XP_011520734.1 |
XM_011522433.2 |
166 |
Intron |
|
|
XP_011520735.1 |
XM_017023079.1 |
166 |
Silent Mutation |
GAA,GAG |
E3E |
XP_016878568.1 |
XM_017023080.1 |
166 |
Silent Mutation |
GAA,GAG |
E3E |
XP_016878569.1 |
XM_017023081.1 |
166 |
Intron |
|
|
XP_016878570.1 |
View Full Product Details