Product Details

SNP ID
rs144487198
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:66599687 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAAGATACTTTTTAAAATGAGGAT[A/G]CTAGCCATTTAGTTCATCCCTAAAG
Phenotype
MIM: 607886 MIM: 607887
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CMTM3 PubMed Links

Gene Details

Gene
CMTM3
Gene Name
CKLF like MARVEL transmembrane domain containing 3
There are no transcripts associated with this gene.

Gene
CMTM4
Gene Name
CKLF like MARVEL transmembrane domain containing 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178818.2 Intron NP_848933.1
NM_181521.2 Intron NP_852662.1
XM_011522882.1 Intron XP_011521184.1
XM_017022954.1 Intron XP_016878443.1

View Full Product Details