Product Details
- SNP ID
-
rs147691988
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:56652603 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTGTGCCTGCGCCGGCTCCTGCAAG[G/T]GCAAAGAGTGCAAATGTACCTCCTG
- Phenotype
-
MIM: 156349
MIM: 156352
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
MT1B
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs12051311,rs964374] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- MT1B
- Gene Name
- metallothionein 1B
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_005947.2 |
116 |
Missense Mutation |
GGC,TGC |
G21C |
NP_005938.1 |
- Gene
- MT1DP
- Gene Name
- metallothionein 1D, pseudogene
There are no transcripts associated with this gene.
- Gene
- MT1F
- Gene Name
- metallothionein 1F
There are no transcripts associated with this gene.
View Full Product Details