Product Details
- SNP ID
-
rs139088043
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:7260394 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCAGGTCACACATACTCCTTGGAAG[A/G]GTTGGACTTAGGGTAAGAGCGGGGT
- Phenotype
-
MIM: 609131
MIM: 610684
MIM: 615019
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CLDN7
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs3744400] are located under a probe and SNP(s) [rs4562] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CLDN7
- Gene Name
- claudin 7
- Gene
- CTDNEP1
- Gene Name
- CTD nuclear envelope phosphatase 1
There are no transcripts associated with this gene.
- Gene
- ELP5
- Gene Name
- elongator acetyltransferase complex subunit 5
View Full Product Details