Product Details

SNP ID
rs140291244
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:81979262 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAGTGAGTTTGCTCCAGCTCAGCA[G/T]CAGGGTCTGAGTATATCTGTGCCCC
Phenotype
MIM: 606236
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ASPSCR1 PubMed Links

Gene Details

Gene
ASPSCR1
Gene Name
ASPSCR1, UBX domain containing tether for SLC2A4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001251888.1 Intron NP_001238817.1
NM_024083.3 Intron NP_076988.1
XM_011523601.2 Intron XP_011521903.1
XM_011523602.2 Intron XP_011521904.1
XM_017025036.1 Intron XP_016880525.1
XM_017025037.1 Intron XP_016880526.1
XM_017025038.1 Intron XP_016880527.1
XM_017025039.1 Intron XP_016880528.1
XM_017025040.1 Intron XP_016880529.1
XM_017025041.1 Intron XP_016880530.1
XM_017025042.1 Intron XP_016880531.1
XM_017025043.1 Intron XP_016880532.1
XM_017025044.1 Intron XP_016880533.1

View Full Product Details