Product Details

SNP ID
rs143871497
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:32444538 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAGCGGTAAGATGGCGGCGGCGGC[G/T]GTGGTGGAGTTCCAGAGAGCCCAGT
Phenotype
MIM: 604449
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
PSMD11 PubMed Links

Gene Details

Gene
PSMD11
Gene Name
proteasome 26S subunit, non-ATPase 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001270482.1 76 Silent Mutation GCG,GCT A5A NP_001257411.1
NM_002815.3 76 Silent Mutation GCG,GCT A5A NP_002806.2

View Full Product Details