Product Details

SNP ID
rs144474638
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:50353942 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGAGATACAGGGAGCGTGGAGGGC[A/G]CCCCCCAGCCCACGGACAATGGCTT
Phenotype
MIM: 608125
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
XYLT2 PubMed Links

Gene Details

Gene
XYLT2
Gene Name
xylosyltransferase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022167.3 871 Missense Mutation ACC,GCC T150A NP_071450.2
XM_005257572.4 871 Missense Mutation ACC,GCC T118A XP_005257629.1

View Full Product Details