Product Details

SNP ID
rs145290029
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:43641920 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTCCATGCAGAATCTCACTCTGAA[C/T]TTGGAGAGGCTGTGGAGTCCCCATC
Phenotype
MIM: 600147
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MEOX1 PubMed Links

Gene Details

Gene
MEOX1
Gene Name
mesenchyme homeobox 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001040002.1 611 Missense Mutation AAT,AGT N137S NP_001035091.1
NM_004527.3 611 Missense Mutation AAT,AGT N252S NP_004518.1
NM_013999.3 611 UTR 3 NP_054705.1
XM_011524818.1 611 Intron XP_011523120.1

View Full Product Details