Product Details
- SNP ID
-
rs145684585
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:81985528 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCAGGTTCGCATCGCTTTGCAGCTG[A/G]ACGATGGCTCGAGGTTGCAGGACTC
- Phenotype
-
MIM: 606236
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ASPSCR1
PubMed Links
Gene Details
- Gene
- ASPSCR1
- Gene Name
- ASPSCR1, UBX domain containing tether for SLC2A4
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001251888.1 |
403 |
Missense Mutation |
AAC,GAC |
N99D |
NP_001238817.1 |
NM_024083.3 |
403 |
Missense Mutation |
AAC,GAC |
N99D |
NP_076988.1 |
XM_011523601.2 |
403 |
Missense Mutation |
AAC,GAC |
N99D |
XP_011521903.1 |
XM_011523602.2 |
403 |
Missense Mutation |
AAC,GAC |
N99D |
XP_011521904.1 |
XM_017025036.1 |
403 |
Missense Mutation |
AAC,GAC |
N99D |
XP_016880525.1 |
XM_017025037.1 |
403 |
Missense Mutation |
AAC,GAC |
N99D |
XP_016880526.1 |
XM_017025038.1 |
403 |
Missense Mutation |
AAC,GAC |
N22D |
XP_016880527.1 |
XM_017025039.1 |
403 |
Missense Mutation |
AAC,GAC |
N22D |
XP_016880528.1 |
XM_017025040.1 |
403 |
Missense Mutation |
AAC,GAC |
N22D |
XP_016880529.1 |
XM_017025041.1 |
403 |
Missense Mutation |
AAC,GAC |
N22D |
XP_016880530.1 |
XM_017025042.1 |
403 |
Missense Mutation |
AAC,GAC |
N99D |
XP_016880531.1 |
XM_017025043.1 |
403 |
Missense Mutation |
AAC,GAC |
N22D |
XP_016880532.1 |
XM_017025044.1 |
403 |
Missense Mutation |
AAC,GAC |
N22D |
XP_016880533.1 |
View Full Product Details