Product Details

SNP ID
rs149386182
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:6447591 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCACAGCCTGAGCCAGAAGAGCAC[C/T]CGGCTGTCTGGAGCCGCCCCTGCCC
Phenotype
MIM: 608921
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FAM64A PubMed Links

Gene Details

Gene
FAM64A
Gene Name
family with sequence similarity 64 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001195228.1 906 Intron NP_001182157.1
NM_019013.2 906 Intron NP_061886.2
XM_011523944.2 906 Silent Mutation ACC,ACT T141T XP_011522246.1
XM_017024778.1 906 Silent Mutation ACC,ACT T95T XP_016880267.1
Gene
PITPNM3
Gene Name
PITPNM family member 3
There are no transcripts associated with this gene.

View Full Product Details