Product Details

SNP ID
rs150029755
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:81562686 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAGTTGGCTGGACATGGTGGCTTA[C/T]CACTGCAATCTCAGCACTGAGGCGG
Phenotype
MIM: 606590
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NPLOC4 PubMed Links

Gene Details

Gene
NPLOC4
Gene Name
NPL4 homolog, ubiquitin recognition factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017921.3 Intron NP_060391.2
XM_011524979.1 Intron XP_011523281.1
XM_011524980.1 Intron XP_011523282.1
XM_011524981.1 Intron XP_011523283.1
XM_011524982.2 Intron XP_011523284.1
XM_017024830.1 Intron XP_016880319.1

View Full Product Details