Product Details

SNP ID
rs150608118
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:82657989 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGTGGTGAGGGAGTAGGAACCGCC[A/G]TGCATCATCCTGGCATTCAGGCCGT
Phenotype
MIM: 609226
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RAB40B PubMed Links

Gene Details

Gene
RAB40B
Gene Name
RAB40B, member RAS oncogene family
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006822.2 949 Silent Mutation CAC,CAT H237H NP_006813.1
XM_005256334.4 949 Silent Mutation CAC,CAT H177H XP_005256391.1
XM_005256335.3 949 Intron XP_005256392.1
XM_006722271.3 949 Silent Mutation CAC,CAT H197H XP_006722334.1
XM_011523528.2 949 Silent Mutation CAC,CAT H220H XP_011521830.1
XM_017024042.1 949 Intron XP_016879531.1
XM_017024043.1 949 Silent Mutation CAC,CAT H177H XP_016879532.1
Gene
WDR45B
Gene Name
WD repeat domain 45B
There are no transcripts associated with this gene.

View Full Product Details