Product Details

SNP ID
rs141174485
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:50282700 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGCGGTTTGTCATGGCTGTGCGCA[C/T]ATTGCGCTCCTGCTCAAACAGCTCG
Phenotype
MIM: 609150 MIM: 156535
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CXXC1 PubMed Links

Gene Details

Gene
CXXC1
Gene Name
CXXC finger protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001101654.1 2342 Missense Mutation ATG,GTG M626V NP_001095124.1
NM_014593.3 2342 Missense Mutation ATG,GTG M622V NP_055408.2
XM_011525940.2 2342 Missense Mutation ATG,GTG M626V XP_011524242.1
XM_011525941.2 2342 Missense Mutation ATG,GTG M501V XP_011524243.1
XM_017025718.1 2342 Missense Mutation ATG,GTG M622V XP_016881207.1
XM_017025719.1 2342 Missense Mutation ATG,GTG M497V XP_016881208.1
Gene
MBD1
Gene Name
methyl-CpG binding domain protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204136.1 2342 Intron NP_001191065.1
NM_001204137.1 2342 Intron NP_001191066.1
NM_001204138.1 2342 Intron NP_001191067.1
NM_001204139.1 2342 Intron NP_001191068.1
NM_001204140.1 2342 Intron NP_001191069.1
NM_001204141.1 2342 Intron NP_001191070.1
NM_001204142.1 2342 Intron NP_001191071.1
NM_001204143.1 2342 Intron NP_001191072.1
NM_001204151.2 2342 Intron NP_001191080.1
NM_001323942.1 2342 Intron NP_001310871.1
NM_001323947.1 2342 Intron NP_001310876.1
NM_001323949.1 2342 Intron NP_001310878.1
NM_001323950.1 2342 Intron NP_001310879.1
NM_001323951.1 2342 Intron NP_001310880.1
NM_001323952.1 2342 Intron NP_001310881.1
NM_001323953.1 2342 Intron NP_001310882.1
NM_001323954.1 2342 Intron NP_001310883.1
NM_002384.2 2342 Intron NP_002375.1
NM_015844.2 2342 Intron NP_056669.2
NM_015845.3 2342 Intron NP_056670.2
NM_015846.3 2342 Intron NP_056671.2
NM_015847.3 2342 Intron NP_056723.2
XM_005258271.2 2342 Intron XP_005258328.1
XM_006722456.2 2342 Intron XP_006722519.1
XM_011525991.1 2342 Intron XP_011524293.1
XM_011525993.2 2342 Intron XP_011524295.1
XM_011525994.2 2342 Intron XP_011524296.1
XM_011525998.2 2342 Intron XP_011524300.1
XM_011525999.1 2342 Intron XP_011524301.1
XM_011526001.1 2342 Intron XP_011524303.1
XM_011526002.1 2342 Intron XP_011524304.1
XM_011526003.2 2342 Intron XP_011524305.1
XM_011526006.1 2342 Intron XP_011524308.1
XM_011526007.1 2342 Intron XP_011524309.1
XM_017025751.1 2342 Intron XP_016881240.1
XM_017025752.1 2342 Intron XP_016881241.1
XM_017025753.1 2342 Intron XP_016881242.1
XM_017025754.1 2342 Intron XP_016881243.1
XM_017025755.1 2342 Intron XP_016881244.1
XM_017025756.1 2342 Intron XP_016881245.1
XM_017025757.1 2342 Intron XP_016881246.1
XM_017025758.1 2342 Intron XP_016881247.1
XM_017025759.1 2342 Intron XP_016881248.1
XM_017025760.1 2342 Intron XP_016881249.1
XM_017025761.1 2342 Intron XP_016881250.1
XM_017025762.1 2342 Intron XP_016881251.1
XM_017025763.1 2342 Intron XP_016881252.1
XM_017025764.1 2342 Intron XP_016881253.1
XM_017025765.1 2342 Intron XP_016881254.1
XM_017025766.1 2342 Intron XP_016881255.1
XM_017025767.1 2342 Intron XP_016881256.1
XM_017025768.1 2342 Intron XP_016881257.1
XM_017025769.1 2342 Intron XP_016881258.1
XM_017025770.1 2342 Intron XP_016881259.1
XM_017025771.1 2342 Intron XP_016881260.1
XM_017025772.1 2342 Intron XP_016881261.1
XM_017025773.1 2342 Intron XP_016881262.1
XM_017025774.1 2342 Intron XP_016881263.1
XM_017025775.1 2342 Intron XP_016881264.1
XM_017025776.1 2342 Intron XP_016881265.1
XM_017025777.1 2342 Intron XP_016881266.1

View Full Product Details