Product Details

SNP ID
rs150531716
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:68715232 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCATGTCTTAAGAATCCTTTGCGCT[C/T]TCGGTGCCCCCCTCCACGCCCAGGA
Phenotype
MIM: 616102
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCDC102B PubMed Links

Gene Details

Gene
CCDC102B
Gene Name
coiled-coil domain containing 102B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001093729.1 11 Intron NP_001087198.1
NM_024781.2 11 Intron NP_079057.2
XM_011526174.2 11 Intron XP_011524476.1
XM_011526175.2 11 Intron XP_011524477.1
XM_017025973.1 11 UTR 5 XP_016881462.1
XM_017025974.1 11 Intron XP_016881463.1
XM_017025975.1 11 UTR 5 XP_016881464.1
XM_017025976.1 11 UTR 5 XP_016881465.1
XM_017025977.1 11 Intron XP_016881466.1
XM_017025978.1 11 Intron XP_016881467.1
XM_017025979.1 11 UTR 5 XP_016881468.1
XM_017025980.1 11 UTR 5 XP_016881469.1
Gene
TMX3
Gene Name
thioredoxin related transmembrane protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019022.3 11 Intron NP_061895.3
XM_005266714.2 11 Intron XP_005266771.1
XM_011526033.2 11 Intron XP_011524335.1
XM_011526034.2 11 Intron XP_011524336.1
XM_011526035.2 11 Intron XP_011524337.1

View Full Product Details