Product Details

SNP ID
rs141077219
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:2757670 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGCCCGCCGCCCACGTCCTCCGTC[A/C]TCTTGGAAGCAGTTCCACTCACGCC
Phenotype
MIM: 603419
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
SGTA PubMed Links

Gene Details

Gene
SGTA
Gene Name
small glutamine rich tetratricopeptide repeat containing alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003021.3 Intron NP_003012.1
XM_011528178.2 Intron XP_011526480.1

View Full Product Details