Product Details

SNP ID
rs141333361
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:10923531 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGCCAGGAGGGCGTGGAGCAGCAC[A/G]ACCACGGACAGCAGCACTGTGGCCA
Phenotype
MIM: 603934
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C19orf52 PubMed Links

Gene Details

Gene
C19orf52
Gene Name
chromosome 19 open reading frame 52
There are no transcripts associated with this gene.

Gene
CARM1
Gene Name
coactivator associated arginine methyltransferase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_199141.1 868 Intron NP_954592.1
XM_005259708.4 868 Intron XP_005259765.1
XM_011527638.2 868 Intron XP_011525940.1
Gene
YIPF2
Gene Name
Yip1 domain family member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321439.1 868 Silent Mutation GTC,GTT V266V NP_001308368.1
NM_001321440.1 868 Silent Mutation GTC,GTT V227V NP_001308369.1
NM_024029.4 868 Silent Mutation GTC,GTT V266V NP_076934.1
XM_011528270.2 868 Silent Mutation GTC,GTT V266V XP_011526572.1
XM_011528271.2 868 Silent Mutation GTC,GTT V237V XP_011526573.1

View Full Product Details