Product Details

SNP ID
rs141973314
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:57506400 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGGCTGAGGCAGCTGCTGAGCAGA[G/T]TGCTTCTGTAGAAGTGCCCAGTTCA
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ZNF773 PubMed Links

Gene Details

Gene
ZNF773
Gene Name
zinc finger protein 773
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304334.1 529 Missense Mutation AGT,ATT S101I NP_001291263.1
NM_001304335.1 529 Intron NP_001291264.1
NM_001304336.1 529 Missense Mutation GTG,TTG V100L NP_001291265.1
NM_001304337.1 529 Intron NP_001291266.1
NM_198542.2 529 Missense Mutation AGT,ATT S102I NP_940944.1

View Full Product Details