Product Details

SNP ID
rs142096783
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.19:42316527 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCATCTACTGGGGTCAATATGCCAC[C/T]GATGGCATTGGCAACGAGAGTGTGA
Phenotype
MIM: 604267
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
MEGF8 PubMed Links

Gene Details

Gene
MEGF8
Gene Name
multiple EGF like domains 8
There are no transcripts associated with this gene.

Gene
PRR19
Gene Name
proline rich 19
There are no transcripts associated with this gene.

Gene
TMEM145
Gene Name
transmembrane protein 145
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173633.2 787 Silent Mutation ACC,ACT T231T NP_775904.2
XM_005258781.4 787 Silent Mutation ACC,ACT T245T XP_005258838.1
XM_011526791.2 787 Silent Mutation ACC,ACT T231T XP_011525093.1
XM_011526792.1 787 Silent Mutation ACC,ACT T245T XP_011525094.1

View Full Product Details