Product Details

SNP ID
rs144033103
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:23653268 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTAGTTTCTCTCCAGTATGTATTTT[G/T]TTATGTTTAGTAAGGTTTGCAGATT
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ZNF675 PubMed Links

Gene Details

Gene
ZNF675
Gene Name
zinc finger protein 675
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138330.2 2465 Intron NP_612203.2
XM_017026442.1 2465 Missense Mutation AAA,AAC K479N XP_016881931.1
XM_017026443.1 2465 Missense Mutation AAA,AAC K479N XP_016881932.1
XM_017026444.1 2465 Intron XP_016881933.1

View Full Product Details