Product Details

SNP ID
rs144649796
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:10115511 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTCGACGTTGAGGATGAGGTGGTC[A/G]TAGCCAAAGCCGGACACCCCGGCAA
Phenotype
MIM: 609336 MIM: 603913 MIM: 602697 MIM: 607793
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ANGPTL6 PubMed Links

Gene Details

Gene
ANGPTL6
Gene Name
angiopoietin like 6
There are no transcripts associated with this gene.

Gene
EIF3G
Gene Name
eukaryotic translation initiation factor 3 subunit G
There are no transcripts associated with this gene.

Gene
P2RY11
Gene Name
purinergic receptor P2Y11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002566.4 Intron NP_002557.2
Gene
PPAN
Gene Name
peter pan homolog (Drosophila)
There are no transcripts associated with this gene.

Gene
PPAN-P2RY11
Gene Name
PPAN-P2RY11 readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001040664.2 Intron NP_001035754.1
NM_001198690.1 Intron NP_001185619.1
Gene
SNORD105
Gene Name
small nucleolar RNA, C/D box 105
There are no transcripts associated with this gene.

Gene
SNORD105B
Gene Name
small nucleolar RNA, C/D box 105B
There are no transcripts associated with this gene.

View Full Product Details