Product Details

SNP ID
rs145667000
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:42302255 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCAGGGACATCCAGCGCCCGTCGC[C/T]CTGTACGTCCTGCACCGGCGTGGGC
Phenotype
MIM: 612082 MIM: 603074
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CIC PubMed Links

Gene Details

Gene
CIC
Gene Name
capicua transcriptional repressor
There are no transcripts associated with this gene.

Gene
PAFAH1B3
Gene Name
platelet activating factor acetylhydrolase 1b catalytic subunit 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145939.1 390 Missense Mutation AGC,GGC S19G NP_001139411.1
NM_001145940.1 390 Missense Mutation AGC,GGC S19G NP_001139412.1
NM_002573.3 390 Missense Mutation AGC,GGC S19G NP_002564.1
XM_017026846.1 390 Missense Mutation AGC,GGC S19G XP_016882335.1
XM_017026847.1 390 Missense Mutation AGC,GGC S19G XP_016882336.1
XM_017026848.1 390 Missense Mutation AGC,GGC S19G XP_016882337.1
Gene
PRR19
Gene Name
proline rich 19
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_199285.2 390 UTR 5 NP_954979.2
XM_005258779.4 390 Intron XP_005258836.2
XM_005258780.3 390 Intron XP_005258837.1
XM_006723154.3 390 Intron XP_006723217.1
XM_011526787.1 390 Intron XP_011525089.1
XM_011526788.1 390 Intron XP_011525090.1
XM_011526789.2 390 Intron XP_011525091.1
XM_011526790.2 390 Intron XP_011525092.1

View Full Product Details