Product Details

SNP ID
rs147207917
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:10923948 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGCCACCGCAGGAAGCCCCACAGG[A/G]CCAGGGGCACCAGCCACGCATAGCA
Phenotype
MIM: 603934
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C19orf52 PubMed Links

Gene Details

Gene
C19orf52
Gene Name
chromosome 19 open reading frame 52
There are no transcripts associated with this gene.

Gene
CARM1
Gene Name
coactivator associated arginine methyltransferase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_199141.1 606 Intron NP_954592.1
XM_005259708.4 606 Intron XP_005259765.1
XM_011527638.2 606 Intron XP_011525940.1
Gene
YIPF2
Gene Name
Yip1 domain family member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321439.1 606 Missense Mutation GCC,GTC A179V NP_001308368.1
NM_001321440.1 606 Missense Mutation GCC,GTC A140V NP_001308369.1
NM_024029.4 606 Missense Mutation GCC,GTC A179V NP_076934.1
XM_011528270.2 606 Missense Mutation GCC,GTC A179V XP_011526572.1
XM_011528271.2 606 Missense Mutation GCC,GTC A150V XP_011526573.1

View Full Product Details