Product Details

SNP ID
rs112240293
Assay Type
Functionally tested
NCBI dbSNP Submissions
11
Location
Chr.1:54983590 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCTGCTGTACCTGGCTTTGGACCA[A/G]GTCCTTCACCTTTTTGGGCTCACTC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TMEM61 PubMed Links

Gene Details

Gene
TMEM61
Gene Name
transmembrane protein 61
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_182532.2 131 Intron NP_872338.1
XM_005270586.4 131 Intron XP_005270643.1
XM_011540911.2 131 Intron XP_011539213.1
XM_011540912.2 131 UTR 5 XP_011539214.1

View Full Product Details