Product Details

SNP ID
rs113018154
Assay Type
Functionally tested
NCBI dbSNP Submissions
10
Location
Chr.1:157515408 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAGCAACCACAGCTGAAGCCCACT[A/G]AGGAATCCAGGAGATGTGCTGGGCC
Phenotype
MIM: 605877
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FCRL5 PubMed Links

Gene Details

Gene
FCRL5
Gene Name
Fc receptor like 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001195388.1 3349 UTR 3 NP_001182317.1
NM_031281.2 3349 UTR 3 NP_112571.2
XM_011510030.1 3349 Intron XP_011508332.1
XM_011510031.1 3349 Intron XP_011508333.1
XM_011510032.1 3349 Intron XP_011508334.1
XM_011510033.1 3349 Intron XP_011508335.1

View Full Product Details