Product Details

SNP ID
rs113133699
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:94983076 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCTAAAGAAGTCAGTTGCCATTTG[C/T]CAAACAATTCGCCCAAGGTACACCG
Phenotype
MIM: 612866
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ALG14 PubMed Links

Gene Details

Gene
ALG14
Gene Name
ALG14, UDP-N-acetylglucosaminyltransferase subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001305242.1 741 UTR 3 NP_001292171.1
XM_005270582.3 741 Intron XP_005270639.1
XM_011540897.2 741 Intron XP_011539199.1

View Full Product Details