Product Details
- SNP ID
-
rs114501214
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
10
- Location
-
Chr.1:204837731 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCAGTGATGCTGCACTGGAAAGTAA[C/T]GCCGGGGAATGCTTACAACCAGCCA
- Phenotype
-
MIM: 609145
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
NFASC
PubMed Links
Gene Details
- Gene
- NFASC
- Gene Name
- neurofascin
View Full Product Details