Product Details

SNP ID
rs115926556
Assay Type
Functionally tested
NCBI dbSNP Submissions
13
Location
Chr.1:99267304 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATGAGGGAAGGGAAACTTACACTC[A/G]AGCCAGAGGGGGCTGAAATCAAGAT
Phenotype
MIM: 607813
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PLPPR4 PubMed Links

Gene Details

Gene
PLPPR4
Gene Name
phospholipid phosphatase related 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001166252.1 Intron NP_001159724.1
NM_014839.4 Intron NP_055654.2
XM_011542498.2 Intron XP_011540800.1

View Full Product Details