Product Details

SNP ID
rs138095068
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:211942017 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCCACTGTAATGTTGTGTGTTCCA[A/G]GGATAGCAAAGTTCAACAGAAATTG
Phenotype
MIM: 611350
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
INTS7 PubMed Links

Gene Details

Gene
INTS7
Gene Name
integrator complex subunit 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199809.1 2877 Missense Mutation CCT,CTT P850L NP_001186738.1
NM_001199811.1 2877 Missense Mutation CCT,CTT P885L NP_001186740.1
NM_001199812.1 2877 Missense Mutation CCT,CTT P879L NP_001186741.1
NM_015434.3 2877 Missense Mutation CCT,CTT P899L NP_056249.1
XM_011509396.2 2877 Missense Mutation CCT,CTT P935L XP_011507698.1
XM_017000962.1 2877 Intron XP_016856451.1

View Full Product Details