Product Details

SNP ID
rs138133260
Assay Type
Functionally tested
NCBI dbSNP Submissions
7
Location
Chr.1:1732461 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTCGGAGATACCTCCTACTCAGGA[C/T]GCAGGAAGCATGGGGCAGGGCAGCG
Phenotype
MIM: 116951
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CDK11A PubMed Links

Gene Details

Gene
CDK11A
Gene Name
cyclin dependent kinase 11A
There are no transcripts associated with this gene.

Gene
SLC35E2
Gene Name
solute carrier family 35 member E2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199787.1 1218 Intron NP_001186716.1
NM_182838.2 1218 Silent Mutation GCA,GCG A265A NP_878258.1

View Full Product Details