Product Details

SNP ID
rs138976951
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:161048240 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCGGACCTGTTGGACCTGAACCAG[A/C]GTGGAAGCTAGACGCATGCCCACGC
Phenotype
MIM: 614264 MIM: 616041 MIM: 191523
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ARHGAP30 PubMed Links

Gene Details

Gene
ARHGAP30
Gene Name
Rho GTPase activating protein 30
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001025598.1 2596 Silent Mutation ACG,ACT T927T NP_001020769.1
NM_001287600.1 2596 Silent Mutation ACG,ACT T779T NP_001274529.1
NM_001287602.1 2596 Silent Mutation ACG,ACT T750T NP_001274531.1
NM_181720.2 2596 Silent Mutation ACG,ACT T716T NP_859071.2
XM_005245070.2 2596 Silent Mutation ACG,ACT T870T XP_005245127.1
XM_005245073.3 2596 Silent Mutation ACG,ACT T779T XP_005245130.1
XM_011509391.2 2596 Silent Mutation ACG,ACT T779T XP_011507693.1
XM_017000960.1 2596 Silent Mutation ACG,ACT T793T XP_016856449.1
Gene
TSTD1
Gene Name
thiosulfate sulfurtransferase like domain containing 1
There are no transcripts associated with this gene.

Gene
USF1
Gene Name
upstream transcription factor 1
There are no transcripts associated with this gene.

View Full Product Details