Product Details
- SNP ID
-
rs139603921
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
4
- Location
-
Chr.1:3021864 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAACCAGAAGAAGTACTTTGTGGGG[A/G]AGGAGGCCCTGTACAAGCAGGAGGC
- Phenotype
-
MIM: 608535
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ACTRT2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs4013154] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ACTRT2
- Gene Name
- actin related protein T2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_080431.4 |
383 |
Missense Mutation |
AAG,GAG |
K60E |
NP_536356.3 |
View Full Product Details