Product Details

SNP ID
rs140018251
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:246727002 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACAGTTGTCCTCAATTGCGTAGGA[C/T]CAGTAAGTAATCAACCCTTCTTTGT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SCCPDH PubMed Links

Gene Details

Gene
SCCPDH
Gene Name
saccharopine dehydrogenase (putative)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016002.2 648 Missense Mutation CCA,TCA P101S NP_057086.2

View Full Product Details