Product Details

SNP ID
rs141244792
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:1309495 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGAGTGCCCAGCGACTTCCACGCTC[A/G]TCACGCAGCCACGTCCCGGACCTAC
Phenotype
MIM: 611354
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ACAP3 PubMed Links

Gene Details

Gene
ACAP3
Gene Name
ArfGAP with coiled-coil, ankyrin repeat and PH domains 3
There are no transcripts associated with this gene.

Gene
CPSF3L
Gene Name
cleavage and polyadenylation specific factor 3-like
There are no transcripts associated with this gene.

Gene
MIR6727
Gene Name
microRNA 6727
There are no transcripts associated with this gene.

Gene
PUSL1
Gene Name
pseudouridylate synthase-like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153339.1 395 Missense Mutation CAT,CGT H122R NP_699170.1
XM_005244720.3 395 Missense Mutation CAT,CGT H122R XP_005244777.1

View Full Product Details