Product Details

SNP ID
rs143155996
Assay Type
Functionally tested
NCBI dbSNP Submissions
7
Location
Chr.1:86424184 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGAAGAAACCATCTGCATCCATA[C/T]TGAAAACCTGACACAATGTATGCAG
Phenotype
MIM: 604003
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CLCA2 PubMed Links

Gene Details

Gene
CLCA2
Gene Name
chloride channel accessory 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006536.5 99 UTR 5 NP_006527.1
XM_011542448.2 99 UTR 5 XP_011540750.1

View Full Product Details