Product Details

SNP ID
rs144284181
Assay Type
Functionally tested
NCBI dbSNP Submissions
6
Location
Chr.1:228457506 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGATGTTGGGCAGGACGCCACCCTG[A/C]GCGATGGTCACGCGGCCCAGCAGCT
Phenotype
MIM: 615015 MIM: 615046
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
HIST3H2A PubMed Links

Gene Details

Gene
HIST3H2A
Gene Name
histone cluster 3, H2a
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033445.2 Intron NP_254280.1
Gene
HIST3H2BB
Gene Name
histone cluster 3, H2bb
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_175055.2 Intron NP_778225.1
Gene
MIR4666A
Gene Name
microRNA 4666a
There are no transcripts associated with this gene.

View Full Product Details