Product Details

SNP ID
rs144741491
Assay Type
Functionally tested
NCBI dbSNP Submissions
13
Location
Chr.1:109559547 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATAAAGGTAACTATTTTTGTGTTA[C/T]GTCTCTTATTCTTGAGTTTGATTGA
Phenotype
MIM: 139370
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GNAI3 PubMed Links

Gene Details

Gene
GNAI3
Gene Name
G protein subunit alpha i3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006496.3 Intron NP_006487.1

View Full Product Details