Product Details

SNP ID
rs145490610
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:44747661 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGATTTGTTTTTCAGAAACAAAAA[C/T]GGAGATCCGTCAACTCCAAAATTCC
Phenotype
MIM: 604538
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KIF2C PubMed Links

Gene Details

Gene
KIF2C
Gene Name
kinesin family member 2C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001297655.1 506 Missense Mutation CGG,TGG R93W NP_001284584.1
NM_001297656.1 506 Missense Mutation CGG,TGG R39W NP_001284585.1
NM_001297657.1 506 UTR 5 NP_001284586.1
NM_006845.3 506 Missense Mutation CGG,TGG R93W NP_006836.2
XM_011540540.2 506 UTR 5 XP_011538842.1
XM_011540541.2 506 UTR 5 XP_011538843.1

View Full Product Details