Product Details

SNP ID
rs145523099
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:147758265 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGGCGGTGACCTGGTGAGACTCCA[C/T]TGGGCACCTCAGGCTTCTGGCCATA
Phenotype
MIM: 121013
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GJA5 PubMed Links

Gene Details

Gene
GJA5
Gene Name
gap junction protein alpha 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005266.6 3157 Missense Mutation AAT,AGT N325S NP_005257.2
NM_181703.3 3157 Missense Mutation AAT,AGT N325S NP_859054.1
XM_005272951.4 3157 Missense Mutation AAT,AGT N325S XP_005273008.1
XM_017001044.1 3157 Missense Mutation AAT,AGT N325S XP_016856533.1

View Full Product Details