Product Details

SNP ID
rs146456804
Assay Type
Functionally tested
NCBI dbSNP Submissions
8
Location
Chr.1:209616391 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATAGCACGGCTAGCTCCAATAAGAA[C/T]GGGCTTCAGAAGCCTTGGGTTGGGT
Phenotype
MIM: 614994 MIM: 150310
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CAMK1G PubMed Links

Gene Details

Gene
CAMK1G
Gene Name
calcium/calmodulin dependent protein kinase IG
There are no transcripts associated with this gene.

Gene
LAMB3
Gene Name
laminin subunit beta 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000228.2 Intron NP_000219.2
NM_001017402.1 Intron NP_001017402.1
NM_001127641.1 Intron NP_001121113.1
XM_005273124.4 Intron XP_005273181.1
XM_017001272.1 Intron XP_016856761.1
Gene
MIR4260
Gene Name
microRNA 4260
There are no transcripts associated with this gene.

View Full Product Details