Product Details
- SNP ID
-
rs147293552
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
5
- Location
-
Chr.1:182646885 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGGATGGCTCCTGCAGGTTCTTCCT[C/T]GTGGCTTCTCGGGTCTGGAAGTCAA
- Phenotype
-
MIM: 607189
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
RGS8
PubMed Links
Gene Details
- Gene
- RGS8
- Gene Name
- regulator of G-protein signaling 8
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001102450.2 |
555 |
Silent Mutation |
ACA,ACG |
T131T |
NP_001095920.1 |
NM_033345.3 |
555 |
Silent Mutation |
ACA,ACG |
T149T |
NP_203131.1 |
XM_005245555.3 |
555 |
Silent Mutation |
ACA,ACG |
T131T |
XP_005245612.1 |
XM_011510089.2 |
555 |
Silent Mutation |
ACA,ACG |
T149T |
XP_011508391.1 |
XM_011510090.2 |
555 |
Silent Mutation |
ACA,ACG |
T131T |
XP_011508392.1 |
XM_011510091.2 |
555 |
Silent Mutation |
ACA,ACG |
T129T |
XP_011508393.1 |
XM_017002631.1 |
555 |
Silent Mutation |
ACA,ACG |
T149T |
XP_016858120.1 |
XM_017002632.1 |
555 |
Silent Mutation |
ACA,ACG |
T149T |
XP_016858121.1 |
XM_017002633.1 |
555 |
Silent Mutation |
ACA,ACG |
T149T |
XP_016858122.1 |
XM_017002634.1 |
555 |
Silent Mutation |
ACA,ACG |
T149T |
XP_016858123.1 |
XM_017002635.1 |
555 |
Silent Mutation |
ACA,ACG |
T131T |
XP_016858124.1 |
XM_017002636.1 |
555 |
Silent Mutation |
ACA,ACG |
T131T |
XP_016858125.1 |
XM_017002637.1 |
555 |
Silent Mutation |
ACA,ACG |
T131T |
XP_016858126.1 |
View Full Product Details