Product Details

SNP ID
rs147529110
Assay Type
Functionally tested
NCBI dbSNP Submissions
14
Location
Chr.1:27966678 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGTCCTGGCTGTGGCCCTGTTCTC[A/G]GCCCTCTTCCCCAGCTATGTGGCCC
Phenotype
MIM: 601655
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EYA3 PubMed Links

Gene Details

Gene
EYA3
Gene Name
EYA transcriptional coactivator and phosphatase 3
There are no transcripts associated with this gene.

Gene
SMPDL3B
Gene Name
sphingomyelin phosphodiesterase acid like 3B
There are no transcripts associated with this gene.

Gene
XKR8
Gene Name
XK related 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018053.3 1274 Silent Mutation TCA,TCG S222S NP_060523.2
XM_011541679.2 1274 Silent Mutation TCA,TCG S276S XP_011539981.1
XM_011541680.2 1274 Silent Mutation TCA,TCG S240S XP_011539982.1

View Full Product Details