Product Details
- SNP ID
-
rs147553599
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
5
- Location
-
Chr.1:156669727 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAGGCTCAGCACTGTCCTGGGACTC[C/T]GTTTCCAGGGCAGTCTTGGGGGCAC
- Phenotype
-
MIM: 600915
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
NES
PubMed Links
Gene Details
- Gene
- NES
- Gene Name
- nestin
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_006617.1 |
4594 |
Silent Mutation |
ACA,ACG |
T1487T |
NP_006608.1 |
View Full Product Details