Product Details

SNP ID
rs148294862
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:158579025 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGGCTGAGGAAGGGGGTAATGACA[A/G]TATAAGGGACTGCTATGAGTGTGTC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR10X1 PubMed Links

Gene Details

Gene
OR10X1
Gene Name
olfactory receptor family 10 subfamily X member 1 (gene/pseudogene)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004477.1 875 Missense Mutation ACT,ATT T292I NP_001004477.1

View Full Product Details