Product Details

SNP ID
rs148693817
Assay Type
Functionally tested
NCBI dbSNP Submissions
8
Location
Chr.1:45693937 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAAAAGTCTGGGAAGTGAGGAGCAC[C/T]TCTGCCCAGCTGCTGCCCCGTCTGG
Phenotype
MIM: 147485
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GPBP1L1 PubMed Links

Gene Details

Gene
GPBP1L1
Gene Name
GC-rich promoter binding protein 1 like 1
There are no transcripts associated with this gene.

Gene
IPP
Gene Name
intracisternal A particle-promoted polypeptide
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145349.1 Intron NP_001138821.1
NM_005897.2 Intron NP_005888.1
XM_005270839.4 Intron XP_005270896.1
XM_006710623.3 Intron XP_006710686.1
XM_011541394.2 Intron XP_011539696.1
XM_017001210.1 Intron XP_016856699.1
Gene
TMEM69
Gene Name
transmembrane protein 69
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016486.3 Intron NP_057570.2

View Full Product Details