Product Details

SNP ID
rs149421007
Assay Type
Functionally tested
NCBI dbSNP Submissions
14
Location
Chr.1:109608665 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGTCATTAAGCTGGTATTCTGCAG[C/T]TCTCTCGAAGCAGGCTTGCACCCCA
Phenotype
MIM: 139340 MIM: 611189
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GNAT2 PubMed Links

Gene Details

Gene
GNAT2
Gene Name
G protein subunit alpha transducin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005272.3 640 Missense Mutation ACT,GCT T143A NP_005263.1
XM_011541264.2 640 Missense Mutation ACT,GCT T143A XP_011539566.1
Gene
MIR197
Gene Name
microRNA 197
There are no transcripts associated with this gene.

View Full Product Details