Product Details

SNP ID
rs150660015
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:159928500 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCTGGAGAACGAAGGAAAGATGAG[A/G]TGGGACAGCGCCGGGTATCCATGTA
Phenotype
MIM: 609738 MIM: 604634
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IGSF9 PubMed Links

Gene Details

Gene
IGSF9
Gene Name
immunoglobulin superfamily member 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135050.1 3086 Missense Mutation NP_001128522.1
NM_020789.3 3086 Missense Mutation NP_065840.2
XM_017001884.1 3086 Intron XP_016857373.1
Gene
TAGLN2
Gene Name
transgelin 2
There are no transcripts associated with this gene.

View Full Product Details