Product Details

SNP ID
rs139774093
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:16273298 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTAGTCAAAGACTCCTTTCTTGAA[A/G]AATGGTGAAAACTCACAAATGGTAT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KIF16B PubMed Links

Gene Details

Gene
KIF16B
Gene Name
kinesin family member 16B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199865.1 3940 Silent Mutation TTC,TTT F1252F NP_001186794.1
NM_001199866.1 3940 Intron NP_001186795.1
NM_024704.4 3940 Silent Mutation TTC,TTT F1303F NP_078980.3
XM_005260750.3 3940 Intron XP_005260807.1
XM_005260751.4 3940 Intron XP_005260808.1
XM_005260752.3 3940 Intron XP_005260809.1
XM_005260753.3 3940 Silent Mutation TTC,TTT F1314F XP_005260810.1
XM_005260754.3 3940 Silent Mutation TTC,TTT F1284F XP_005260811.1
XM_005260755.3 3940 Silent Mutation TTC,TTT F1263F XP_005260812.1
XM_006723588.3 3940 Silent Mutation TTC,TTT F1273F XP_006723651.1
XM_017027926.1 3940 Silent Mutation TTC,TTT F1262F XP_016883415.1

View Full Product Details