Product Details

SNP ID
rs143738831
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:58986147 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGATGCGGAGGTTCAGCAAGAATGC[C/T]TGCATAAATTTTCCACCCGGGATTA
Phenotype
MIM: 603169 MIM: 605297
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CTSZ PubMed Links

Gene Details

Gene
CTSZ
Gene Name
cathepsin Z
There are no transcripts associated with this gene.

Gene
NELFCD
Gene Name
negative elongation factor complex member C/D
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198976.2 217 Silent Mutation CTG,TTG L57L NP_945327.2

View Full Product Details