Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001317012.1 | 3124 | Intron | NP_001303941.1 | ||
NM_001317019.1 | 3124 | Intron | NP_001303948.1 | ||
NM_022482.4 | 3124 | Intron | NP_071927.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001283018.1 | 3124 | UTR 3 | NP_001269947.1 | ||
NM_001283020.1 | 3124 | UTR 3 | NP_001269949.1 | ||
NM_001283026.1 | 3124 | UTR 3 | NP_001269955.1 | ||
NM_022080.2 | 3124 | UTR 3 | NP_071363.1 | ||
XM_011529313.1 | 3124 | UTR 3 | XP_011527615.1 | ||
XM_011529315.2 | 3124 | Intron | XP_011527617.1 | ||
XM_017028008.1 | 3124 | UTR 3 | XP_016883497.1 |